Europium(III) perchlorate hexahydrate 50% (w/w) in aqueous solution, Reagent Grade

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: Europium(III) perchlorate

40572-NC 40572-NC
AA40572-NCEA 383 USD
CAAA40572-NC AA40572-NC
Europium(III) perchlorate hexahydrate 50% (w/w) in aqueous solution, Reagent Grade
Europium (III) perchlorate hexahydrate
Formula: Cl₃EuO₁₂·6H₂O
MW: 558.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150233
CAS Number: 36907-40-1
EINECS: 236-907-5
UN: 3211
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR