Tetrakis(diethylamino)titanium (TDEAT) ≥99.99% (metals basis)

Danger

Synonyms: Tetrakis(diethylamino)titanium(IV)

40469-18 40469-09
CAAA40469-09 CAAA40469-18
Tetrakis(diethylamino)titanium (TDEAT) ≥99.99% (metals basis)
Tetrakis(diethylamino)titanium (TDEAT)

Applications: Nitride precursor - MOCVD

Formula: C₁₆H₄₀N₄Ti
MW: 336.383  g/mol
Boiling Pt: 133 °C (1.2 mmHg)
Density: 0.95
Storage Temperature: Ambient
MDL Number: MFCD00015648
EINECS: 437-500-7
UN: 3129
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR