Pentakis(diethylamino)niobium(V) mixture of complexes ≥99.9% (metals basis)

Danger

40445-27 40445-27 40445-ME 40445-ME
AA40445-MEEA 83.77 USD
AA40445-ME CAAA40445-ME AA40445-27 CAAA40445-27
Pentakis(diethylamino)niobium(V) mixture of complexes ≥99.9% (metals basis)
Pentakis(diethylamino)niobium(V)

Applications: Nitride film precursor

Formula: C₂₀H₅₀N₅Nb
Boiling Pt: 120 °C (0.1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00798536
CAS Number: 25169-05-5
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR