Cesium metasilicate ≥99%

Supplier: Thermo Scientific Chemicals
Warning

40208-14 040208-14
AA40208-14EA 230.99 USD
AA40208-14 CAAA40208-14
Cesium metasilicate ≥99%
Cesium metasilicate
Formula: Cs₂O₃Si
Storage Temperature: Ambient
MDL Number: MFCD00798519
CAS Number: 15586-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR