Tin(II) phthalocyanine
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tin(ii)phthalocyanine
39607-02
039607-02
AA39607-02EA
68.59
USD
AA39607-02
CAAA39607-02
Tin(II) phthalocyanine
Tin (II) phthalocyanine
Formula:
C₃₂H₁₆N₈Sn MW: 631.233 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00058867 CAS Number: 15304-57-1 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...