Tris[N,N-bis(trimethylsilyl)amide]yttrium(III) 98+%

Danger

039578-03 39578-03
AA39578-03EA 128.75 USD
AA39578-03 CAAA39578-03
Tris[N,N-bis(trimethylsilyl)amide]yttrium(III) 98+%
Tris[N,N-bis(trimethylsilyl)amide]yttrium (III)
Formula: C₁₈H₅₄N₃Si₆Y
MW: 570.06 g/mol
Boiling Pt: 105 °C (10^-4 mmHg) (subl.)
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00210649
CAS Number: 41836-28-6
UN: 3396
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR