Diethylzinc ≥96%

Supplier: Thermo Scientific Chemicals
Danger

039564-22 39564-22
AA39564-22EA 317.99 USD
AA39564-22 CAAA39564-22
Diethylzinc ≥96%
Diethylzinc
Formula: C₄H₁₀Zn
Boiling Pt: 124 °C
Melting Pt: –28 °C
Density: 1,18
Flash Pt: –18 °C (–1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009021
CAS Number: 557-20-0
EINECS: 209-161-3
UN: 3394
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR