Bis(pentamethylcyclopentadienyl)zirconium(IV) dichloride 99%

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: Bis(pentamethylcyclopentadienyl)zirconium dichloride

39555-04 39555-04
AA39555-04EA 244 USD
CAAA39555-04 AA39555-04
Bis(pentamethylcyclopentadienyl)zirconium(IV) dichloride 99%
Bis(pentamethylcyclopentadienyl)zirconium(IV) dichloride
Formula: C₂₀H₃₀Cl₂Zr
MW: 432.582  g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058849
CAS Number: 54039-38-2
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR