Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate

Synonyms: Bis(1,5-cyclooctadiene)iridium (I) tetrafluoroborate

39414-99 39414-ME 039414-ME 39414-03 39414-99 39414-03
AAA39414-99EA 2193.45 USD
AAA39414-99 AA39414-03 CAAA39414-03 CAAA39414-ME CAAAA39414-99 AA39414-ME
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Bis(1,5-cyclooctadiene)iridium (I) tetrafluoroborate

Soluble in chloroform, methylene chloride

Formula: C₁₆H₂₄BF₄Ir
MW: 495.383 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079664
CAS Number: 35138-23-9

Order Now

Specification Test Results

Form Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR