Carbonylchlorobis(triphenylphosphine)iridium(I)

Danger

Synonyms: Chlorocarbonylbis(triphenylphosphine)iridium(I)VASKA'S COMPLEXCarbonylchloro bis(triphenylphosphine)iridium(I)

039413-01 39413-06 39413-03 39413-01 039413-03
AA39413-01EA 72.19 USD
AA39413-01 CAAA39413-03 CAAA39413-01 CAAA39413-06 AA39413-03
Carbonylchlorobis(triphenylphosphine)iridium(I)
Carbonylchlorobis(triphenylphosphine)iridium(I)

Soluble in chloroform, toluene. Slightly soluble in acetone, alcohol

Formula: C₃₇H₃₀ClIrOP₂
MW: 780.26 g/mol
Melting Pt: 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009590
CAS Number: 14871-41-1
EINECS: 238-941-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR