Chlorobis(ethylene)rhodium(I) dimer ≥99%

Synonyms: Dichlorotetra(ethylene)dirhodium(I)µ-Dichlorotetraethylene dirhodium(I)Bis(ethylene)rhodium(I) chloride dimer

039291-03 39291-03
AA39291-03EA 667.99 USD
CAAA39291-03 AA39291-03
Chlorobis(ethylene)rhodium(I) dimer ≥99%
Chlorobis(ethylene)rhodium(I) dimer
Formula: C₈H₁₆Cl₂Rh₂
MW: 388.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013206
CAS Number: 12081-16-2
EINECS: 235-145-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR