Chlorophenol red sodium salt 0,04% (w/v) in aqueous solution

38699-AP 38699-M0 38699-AP 38699-AE 38699-M0 38699-AE
AA38699-AEEA 31.45 USD
AA38699-AE AA38699-AP CAAA38699-AP CAAA38699-AE AA38699-M0 CAAA38699-M0
Chlorophenol red sodium salt 0,04% (w/v) in aqueous solution
Chlorophenol red sodium salt

Transition interval: pH 4.8 (yellow) to 6.4 (red)

Formula: C₁₉H₁₂Cl₂O₅S
Density: 0,990
Storage Temperature: Ambient
MDL Number: MFCD00151199
CAS Number: 123333-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR