Bis(ethylcyclopentadienyl)magnesium 98+%

Danger

38597-MD 38597-MD 38597-03 38597-06 38597-06 38597-03
AA38597-06EA 700.92 USD
CAAA38597-MD AA38597-06 AA38597-MD CAAA38597-03 AA38597-03 CAAA38597-06
Bis(ethylcyclopentadienyl)magnesium 98+%
Bis(ethylcyclopentadienyl)magnesium
Formula: C₁₄H₁₈Mg
Density: 1,91
Storage Temperature: Ambient
MDL Number: MFCD00151375
CAS Number: 114460-02-5
UN: 3392
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR