Lithium hexafluoroantimonate 97%

Supplier: Thermo Scientific Chemicals

Warning

38595-14 38595-14
AA38595-14EA 464.47 USD
CAAA38595-14 AA38595-14
Lithium hexafluoroantimonate 97%
Lithium hexafluoroantimonate

Very soluble in water

Formula: F₆LiSb
MW: 242.68
Boiling Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00042319
CAS Number: 18424-17-4
UN: 1549
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR