Tris(cyclopentadienyl)gadolinium(III) ≥99.9% (REO, rare earth oxide basis)
|
Warning
|
38585-06
38585-06
38585-03
38585-03
AA38585-06EA
668.97
USD
CAAA38585-06
AA38585-06
CAAA38585-03
AA38585-03
Tris(cyclopentadienyl)gadolinium(III) ≥99.9% (REO, rare earth oxide basis)
Tris(cyclopentadienyl)gadolinium(III)
Formula:
C₁₅H₁₅Gd Storage Temperature: Ambient |
MDL Number:
MFCD00145476 CAS Number: 1272-21-5 EINECS: 215-046-9 UN: 3181 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...