Dinickel silicide ≥99% (metals basis excluding Co) (max. 0.1-1% Co)

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: Nickel silicide

36251-18 36251-30 36251-18 36251-30
AA36251-30EA 306 USD
CAAA36251-18 AA36251-30 CAAA36251-30 AA36251-18
Dinickel silicide ≥99% (metals basis excluding Co) (max. 0.1-1% Co)
Dinickel silicide
Formula: Ni₂Si
Storage Temperature: Ambient
MDL Number: MFCD00151367
CAS Number: 12059-14-2
EINECS: 235-033-1
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR