(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer dimer ≥99%

Danger

Synonyms: Dichloro(pentamethylcyclopentadienyl)rhodium(III) Dimer

33657-03 033657-MD 33657-MD 033657-03
AA33657-03EA 780.99 USD
AA33657-03 CAAA33657-MD AA33657-MD CAAA33657-03
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer dimer ≥99%
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer
Formula: C₂₀H₃₀Cl₄Rh₂
MW: 618.07 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00061552
CAS Number: 12354-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR