Dicyclohexylchlorophosphine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chlorodicyclohexylphosphine
033571-06
33571-03
033571-03
33571-06
AA33571-06EA
631.99
USD
CAAA33571-03
AA33571-06
CAAA33571-06
AA33571-03
Dicyclohexylchlorophosphine 98+%
Dicyclohexylchlorophosphine
Formula:
C₁₂H₂₂ClP Boiling Pt: 97 °C (0.1 mmHg) Density: 1.054 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014294 CAS Number: 16523-54-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...