Hexafluoroantimonic acid ∼65% in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hydrogen hexafluoroantimonate(V)
33488-14
33488-22
33488-22
033488-14
AA33488-22EA
371.76
USD
CAAA33488-14
AA33488-22
AA33488-14
CAAA33488-22
Hexafluoroantimonic acid ∼65% in aqueous solution
Hexafluoroantimonic acid
Formula:
HF₆Sb MW: 236.758 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011327 CAS Number: 16950-06-4 EINECS: 241-023-8 UN: 3264 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...