Ceramic Tubes, AL23, Both Ends Open, Thermo Scientific
Supplier: Thermo Scientific
31555-EO 31560-EH 31558-QAT 031558-DW 31558-DR 31555-EH 31560-DR 31556-DW 31562-DM 31556-EO 31556-QAT 31562-DW 31556-DM 31563-DM 031562-QAT 31558-DR 31553-DR 31558-DW 31553-DR 031563.DW 31558-DM 31555-EM 31555-EO 031563.EM 31560-EH 31562-EH 31560-DW 031554-QAT 31553-EH 31553-EO 31558-EH 031560-QAT 31560-DM 31560-DM 31553-QAT 31555-EH 031556-QAT 31556-EM 31562-DR 31556-DM 31556-DR 31556-EH 31555-EM 031562.EO 031562.DW 31562-DM 031558-DM 031554-DW 31562-QAT 31562-EO 31563-DM 31554-DR 31553-DM 31555-QAT 031558-EO 31563-EO 031554-DR 31554-DM 31555-DM 031560.EM 031563.DR 31553-DW 31560-QAT 31553-DM 31563-DR 31563-EH 31555-DR 31562-EM 031563-QAT 31560-DR 31555-DW 31558-EM 031554-EM 31553-QAT 31556-DR 31560-EM 31553-EH 31553-EM 31562-DR 31555-QAT 31554-EH 31560-EO 31553-EO 31554-EO 31558-QAT 31555-DW 31554-EO 31558-EM 31556-EO 31556-EH 31562-EM 31562-EH 31554-DM 031554-EH 31554-EM 31558-EH 31556-EM 31555-DR 31554-QAT 31555-DM 031563.EH 31553-EM 31558-EO 031563.EO 31554-DW 31563-QAT 31560-EO 31563-DW 31553-DW 31556-DW 31563-EM 31560-DW
AA31554-DREA 41.59 USD
AA31554-DR CAAA31562-DR CAAA31562-DM AA31554-DM CAAA31553-EM CAAA31556-DM AA31560-EH AA31554-DW CAAA31562-DW CAAA31553-EH CAAA31553-DR AA31560-QAT AA31560-DM AA31554-EM AA31560-DR AA31560-DW AA31554-EH CAAA31553-DW CAAA31562-EH CAAA31558-EM CAAA31558-EO CAAA31562-EM CAAA31562-EO CAAA31553-DM CAAA31558-EH CAAA31558-QAT AA31562-EO CAAA31556-QAT AA31562-EM CAAA31558-DW CAAA31560-DM CAAA31555-EH AA31555-QAT AA31556-DW CAAA31558-DM CAAA31555-QAT AA31553-EO AA31553-EM CAAA31558-DR CAAA31555-EO AA31562-EH CAAA31560-DR CAAA31555-EM AA31556-DR AA31563-QAT CAAA31560-DW CAAA31555-DW AA31556-EH AA31556-EM CAAA31555-DR AA31562-DM AA31556-EO AA31562-DW AA31562-DR CAAA31560-EH AA31553-DR CAAA31563-DR CAAA31560-EM CAAA31563-DM AA31553-DM CAAA31560-EO CAAA31555-DM AA31553-DW CAAA31563-DW AA31556-DM CAAA31553-QAT CAAA31563-EM AA31553-EH CAAA31563-EH CAAA31563-EO AA31555-EO AA31558-DR AA31558-DW AA31558-EM AA31558-EO AA31558-EH AA31555-DR CAAA31560-QAT CAAA31554-EO CAAA31554-EM AA31555-DW CAAA31554-EH CAAA31562-QAT AA31558-DM CAAA31554-DR AA31555-EM AA31555-EH AA31553-QAT CAAA31554-DW CAAA31563-QAT CAAA31554-DM CAAA31554-QAT AA31554-QAT AA31556-QAT AA31563-EM AA31563-EO AA31555-DM CAAA31556-EH AA31554-EO AA31563-EH AA31558-QAT CAAA31556-EO CAAA31556-EM CAAA31556-DW AA31562-QAT AA31563-DR AA31560-EO CAAA31553-EO CAAA31556-DR AA31563-DM AA31560-EM AA31563-DW
Ceramic Tubes, AL23, Both Ends Open, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL23.
  • Corrosion resistant
  • Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.
Outer diameter ranges from 3 to 11 mm.