1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4,8,11-Tetramethylcyclam

030854-03 30854-03
AA30854-03EA 282 USD
AA30854-03 CAAA30854-03
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane
Formula: C₁₄H₃₂N₄
MW: 256.44 g/mol
Melting Pt: 34…36 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005106
CAS Number: 41203-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR