Ceramic Tubes, AL23, One End Closed, Thermo Scientific
Supplier: Thermo Scientific
030072-DR 30072-DM 30073-DM 30072-QAT 30066-DM 030074-DR 30069-EM 30073-EO 030059-EH 30073-EH 30066-DW 30072-DM 30062-DW 30074-DR 30059-DW 030074-DM 30074-EH 30059-EH 030066-DR 030062-DM 030074-EH 30073-EH 030059-QAT 030064-DM 30069-DM 30072-EM 30066-QAT 30069-DW 30074-EM 30073-QAT 30072-DR 30066-EM 030066-DM 30072-EM 30064-DR 030059-DM 30074-DW 30064-EO 30069-EH 30064-DR 30072-EO 30069-EH 30072-EH 30059-QAT 30062-DM 30066-DW 30064-EH 030059-DR 30062-DR 030069-QAT 030059-EO 30074-EO 030069-DR 030062-DR 30062-EM 30066-EO 30073-DM 030066-EM 30074-QAT 30064-EM 30069-EM 030062-QAT 30059-DR 30066-DR 30073-DR 30064-DW 030073-QAT 30064-QAT 30059-EO 030059-DW 030062-DW 30064-EH 30069-DR 30062-QAT 30064-QAT 30059-EM 30062-EH 030062-EO 030073-DR 30073-DW 30064-EM 30059-EM 30072-DW 30073-DW 30059-DM 30072-EH 30074-DM 30069-EO 30064-EO 030074-DW 030062-EH 30069-EO 30062-EO 30074-EM 30074-QAT 30064-DW 30069-QAT 30069-DM 30066-EH 030062-EM 30074-EO 30072-QAT 30069-DW 30072-DW 30073-EM 30066-QAT 30064-DM 030073-EM 30066-EO 30072-EO 30066-EH 30073-EO
AA30059-EHEA 70.99 USD
CAAA30073-EM CAAA30073-EO AA30059-EH AA30074-DW AA30059-EM AA30072-QAT AA30074-DM AA30059-DW CAAA30064-EO CAAA30064-EM AA30064-QAT AA30074-DR AA30059-DR CAAA30073-DM AA30059-DM CAAA30073-QAT CAAA30062-EH CAAA30073-DW CAAA30073-DR CAAA30062-DM AA30074-EM AA30074-EO AA30074-EH CAAA30059-DM CAAA30062-DW CAAA30073-EH AA30066-QAT CAAA30062-DR CAAA30074-DM AA30066-DM AA30072-DW CAAA30059-DR CAAA30074-DW AA30066-DW CAAA30074-DR CAAA30059-DW AA30066-DR CAAA30062-EM CAAA30062-EO CAAA30059-EO AA30069-QAT CAAA30059-EM AA30072-DR CAAA30074-EH AA30066-EH AA30072-DM CAAA30059-QAT CAAA30059-EH AA30069-DW AA30066-EO AA30066-EM AA30069-DM AA30069-DR AA30072-EM AA30069-EH AA30069-EM AA30072-EH AA30072-EO AA30062-QAT AA30064-DM CAAA30072-DM CAAA30072-DR AA30069-EO AA30064-DW AA30064-DR CAAA30064-QAT CAAA30072-DW CAAA30072-QAT AA30064-EH AA30073-QAT AA30064-EO CAAA30074-EO AA30064-EM AA30073-DW CAAA30069-DM CAAA30074-QAT CAAA30074-EM CAAA30066-EH CAAA30069-QAT CAAA30066-DR AA30073-DM CAAA30066-DM CAAA30062-QAT AA30073-DR CAAA30066-DW CAAA30069-EH AA30062-DM CAAA30069-EM CAAA30069-EO AA30059-QAT AA30062-DR CAAA30069-DW AA30073-EM AA30062-DW AA30073-EH CAAA30066-EO CAAA30066-EM CAAA30069-DR AA30073-EO CAAA30072-EH AA30062-EM CAAA30072-EO AA30062-EH CAAA30072-EM CAAA30064-EH AA30062-EO AA30074-QAT CAAA30064-DM CAAA30066-QAT AA30059-EO CAAA30064-DW CAAA30064-DR
Ceramic Tubes, AL23, One End Closed, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL23.
  • Corrosion resistant
  • Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.
Outer diameter ranges from 3 to 15 mm.