Cyclopentadienylzirconium trichloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

20617-MD 20617-MD 20617-06 20617.06
AA20617-06EA 247.99 USD
AA20617-06 CAAA20617-MD AA20617-MD CAAA20617-06
Cyclopentadienylzirconium trichloride 98+%
Cyclopentadienylzirconium trichloride
Formula: C₅H₅Cl₃Zr
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00015756
CAS Number: 34767-44-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR