Dysprosium(III) oxalate decahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Warning

14593-14 14593-22 14593-22 14593-14
AA14593-14EA 63.9 USD
CAAA14593-22 AA14593-14 CAAA14593-14 AA14593-22
Dysprosium(III) oxalate decahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Dysprosium (III) oxalate decahydrate
Formula: C₆Dy₂O₁₂·10H₂O
MW: 769.21 (589.06 anhy)
Melting Pt: 40 °C (-H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00150482
CAS Number: 24670-07-3
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR