Yttrium(III) oxalate nonahydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

014562-18 14562-09 14562-18
AA14562-18EA 116.99 USD
CAAA14562-18 AA14562-18 AA14562-09
Yttrium(III) oxalate nonahydrate ≥99.9% (REO, rare earth oxide basis)
Yttrium(III) oxalate nonahydrate
Formula: C₆O₁₂Y₂·9H₂O
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00150502
CAS Number: 13266-82-5
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR