Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate ≥98%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium hydrate[Ba(TMHD)2]

14539-06 14539-03 14539-06 14539-03
AA14539-03EA 56.77 USD
CAAA14539-06 AA14539-03 AA14539-06 CAAA14539-03
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate ≥98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate

Applications: MOCVD of superconductors

Formula: C₂₂H₃₈BaO₄·H₂O
MW: 521.877 g/mol
Boiling Pt: 225 °C (0.3 mmHg)
Melting Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00058709
CAS Number: 17594-47-7
UN: 1564
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR