Barium hexafluoroacetylacetonate

Warning

Synonyms: Barium hexafluoro-2,4-pentanedionate

14503-06 14503-06 14503-03 14503-03
AA14503-03EA 35.96 USD
CAAA14503-03 AA14503-03 CAAA14503-06 AA14503-06
Barium hexafluoroacetylacetonate
Barium hexafluoroacetylacetonate
Formula: C₁₀H₂BaF₁₂O₄
MW: 551.44 g/mol
Melting Pt: 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00075633
CAS Number: 118131-57-0
UN: 1564
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR