Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III) 98+%

Warning

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III)Y(TMHD)3

14480-06 14480-06 14480-03 14480-03
AA14480-06EA 357.58 USD
AA14480-06 CAAA14480-06 AA14480-03 CAAA14480-03
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III) 98+%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III)
Formula: C₃₃H₅₇O₆Y
MW: 638.708 g/mol
Boiling Pt: 95 °C (0.05 mmHg)
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00015713
CAS Number: 15632-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR