Brass (alloy 260), foil 15x45 cm (5.9x18 in), Thickness 0.51 mm (0.02 in)
13502-KJ 13502-KG 13502-KJ 13502-KG
AA13502-KGEA 70.49 USD
CAAA13502-KG CAAA13502-KJ AA13502-KG AA13502-KJ
Brass (alloy 260), foil 15x45 cm (5.9x18 in), Thickness 0.51 mm (0.02 in)
Brass (alloy 260)
Typical composition of alloy: Cu 68.5-71.5%, Pb 0.07% max, Fe 0.05% max, Zn remainder