Erbium(III) oxalate decahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Warning

13216-09 13216-09
AA13216-09EA 55.26 USD
CAAA13216-09 AA13216-09
Erbium(III) oxalate decahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Erbium (III) oxalate decahydrate
Formula: C₆Er₂O₁₂·10H₂O
MW: 778.73 (598.58 anhy)
Density: 2.64
Storage Temperature: Ambient
MDL Number: MFCD00150142
CAS Number: 30618-31-6
EINECS: 212-764-4
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR