Nickel boride ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
13110.22
13110-22
AA13110-22EA
305.99
USD
AA13110-22
CAAA13110-22
Nickel boride ≥99% (metals basis)
Nickel boride
Formula:
BNi Density: 7,39 Storage Temperature: Ambient |
MDL Number:
MFCD00016261 CAS Number: 12007-00-0 EINECS: 234-493-0 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...