Potassium acetate hydrate ≥99.997% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

12975.06 12975-06 12975.14 12975-14
AA12975-14EA 300.99 USD
CAAA12975-06 AA12975-14 CAAA12975-14 AA12975-06
Potassium acetate hydrate ≥99.997% (metals basis), Puratronic®
Potassium acetate hydrate
Formula: C₂H₃KO₂·H₂O
MW: 98.15 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 254966-93-3
EINECS: 204-822-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR