Zinc selenite

Danger

12586-09 12586-A3 12586-22 012586-22 12586-36 012586-36 012586-09 012586-A3
AA12586-09EA 17.39 USD
AA12586-09 CAAA12586-09 AA12586-36 CAAA12586-36 AA12586-A3 CAAA12586-22 AA12586-22 CAAA12586-A3
Zinc selenite
Zinc selenite
Formula: O₃SeZn
MW: 192.33
Storage Temperature: Ambient
MDL Number: MFCD00011297
CAS Number: 13597-46-1
EINECS: 237-048-9
UN: 2630
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR