Calcium perchlorate hydrate (12.2 - 13.5% Ca), Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

11655-30 11655.3
AA11655-30EA 64.19 USD
CAAA11655-30 AA11655-30
Calcium perchlorate hydrate (12.2 - 13.5% Ca), Reagent Grade
Calcium perchlorate hydrate
Formula: CaClO₄·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00015971
CAS Number: 13477-36-6
EINECS: 236-768-0
UN: 1455
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR