Lithium thiocyanate hydrate

Supplier: Thermo Scientific Chemicals
Warning

11349-18 11349-18
AA11349-18EA 155.98 USD
AA11349-18 CAAA11349-18
Lithium thiocyanate hydrate
Lithium thiocyanate hydrate
Formula: LiSCN•xH2O
MW: 65.02 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149773
CAS Number: 123333-85-7
EINECS: 209-135-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR