Potassium tetracyanopalladate(II) hydrate

Supplier: Thermo Scientific Chemicals
Warning

010535-03 10535-03
AA10535-03EA 198.99 USD
CAAA10535-03 AA10535-03
Potassium tetracyanopalladate(II) hydrate
Potassium tetracyanopalladate(II) hydrate
Formula: K2[Pd(CN)4]•xH2O
MW: 288.689 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150431
CAS Number: 14516-46-2
EINECS: 238-520-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR