Diethylphenylphosphine 97%

Danger

Synonyms: Diethyl(phenyl)phosphane

010180-14 10180-14
AA10180-14EA 567.53 USD
AA10180-14 CAAA10180-14
Diethylphenylphosphine 97%
Diethylphenylphosphine
Formula: C₁₀H₁₅P
MW: 166.21 g/mol
Boiling Pt: 220 °C
Density: 0.954
Flash Pt: 66 °C (150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015172
CAS Number: 1605-53-4
EINECS: 216-516-6
UN: 3278
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR