Interceptor Class II Biological Safety Cabinets, Kewaunee Scientific
Supplier: KEWAUNEE SCIENTIFIC
INT-2000A-1 INT1100A21-230 INT1100A2-230 INT-2000B INT1100A2 INT1100A2-1 INT-1700A INT-1400A-1 INT-1400A INT-2000A INT-1700A-1 INT-1100A-1 INT-1100A INT-2000B-1 INT-1400B-1 INT-1400B
89369-244EA 32747.15 USD InStock
89369-244 76208-056 76208-158 89369-246 76208-156 76208-160 89369-234 89369-224 89369-222 89369-242 89369-236 89369-216 89369-214 89369-248 89369-228 89369-226
Interceptor Class II Biological Safety Cabinets, Kewaunee Scientific
Fume Hoods and Enclosures Biological and Microbiological Safety Cabinets
The Interceptor Class II, biological safety cabinets are Kewaunee’s latest design
Ergonomic design includes 10 degree angled easy lift sash utilizing a reinforced nylon sash belt and sprocket. Easy access, touch pad controls ensuring ADA compliance. Removable single piece stainless steel interior worksurface. Single piece interior liner features coved corners for ease of cleaning. Forward positioned service fittings ensure user can easily reach for both ergonomics and safety. All Interceptor biological safety cabinets come standard with base stand and ergonomic arm rest. Choice of electrically or mechanically adjustable stand. Base stand range of motion is 28 to 33".
Kewaunee's full color VGA monitor, with high resolution 640 x 480 pixel display, offers superior clarity. Fully programmable screen ensures operator safety by displaying continuous "Face Velocity" and "Remaining Filter Capacity" readouts at all times. In addition to all alarm features, the VGA monitor also features a pre-programmed user training demonstration.
Developed with comprehensive features created to enhance both safety and ergonomics. Standard features include full color VGA monitor with real time performance data. Multiple audible and visual alarms features include sash position alarm, programmable filter load warning, maximum filter load alarm, UV lamp "ON" indicator, UV lamp life expectancy monitor, and low airflow sensor (thimble connection).