2-Bromo-3-quinolineboronic acid 97%

Warning

L20327-MD L20327-03 L20327-MD L20327-03 L20327-06 L20327-06
AAAL20327-03EA 223.11 USD
CAAAAL20327-03 CAAAAL20327-06 CAAAAL20327-MD AAAL20327-03 AAAL20327-06 AAAL20327-MD
2-Bromo-3-quinolineboronic acid 97%
2-Bromo-3-quinolineboronic acid
Formula: C₉H₇BBrNO₂
Melting Pt: 144…147 °C
Storage Temperature: Ambient
MDL Number: MFCD05664125
CAS Number: 745784-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR