3-(Methylthio)phenylboronic acid 97%

Warning

Synonyms: 3-(Methylthio)benzeneboronic acid3-Thioanisoleboronic acid

L20250-06 L20250-MD L20250-03 L20250-MD L20250-06 L20250-03
AAAL20250-03EA 92.87 USD
AAAL20250-03 CAAAAL20250-06 AAAL20250-06 CAAAAL20250-MD CAAAAL20250-03 AAAL20250-MD
3-(Methylthio)phenylboronic acid 97%
3-(Methylthio)phenylboronic acid
Formula: C₇H₉BO₂S
MW: 168.02 g/mol
Melting Pt: 158…161 °C
Storage Temperature: Ambient
MDL Number: MFCD01319105
CAS Number: 128312-11-8
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR