5-Bromo-3-pyridineboronic acid tech. 85%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromopyridyl-5-boronic acid3-Bromopyridine-5-boronic acid

L20084-MD L20084-06 L20084-03 L20084-03
AAAL20084-03EA 153.99 USD
CAAAAL20084-MD CAAAAL20084-06 AAAL20084-03 CAAAAL20084-03
5-Bromo-3-pyridineboronic acid tech. 85%, Technical Grade
5-Bromo-3-pyridineboronic acid
Formula: C₅H₅BBrNO₂
MW: 201.81 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02685634
CAS Number: 452972-09-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR