2,3,5-Trifluoroisonicotinic acid 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 2,3,5-Trifluoropyridine-4-carboxylic acid

L20050-06 L20050-06 L20050-MD L20050-03 L20050-03
AAAL20050-06EA 413 USD
AAAL20050-06 CAAAAL20050-MD CAAAAL20050-06 AAAL20050-03 CAAAAL20050-03
2,3,5-Trifluoroisonicotinic acid 97%
2,3,5-Trifluoroisonicotinic acid
Formula: C₆H₂F₃NO₂
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD03840205
CAS Number: 675602-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR