2-Acetamido-5-aminopyridine 98%

Warning

L20004-03 L20004-03 L20004-06 L20004-06
AAAL20004-06EA 148.32 USD
AAAL20004-06 CAAAAL20004-06 AAAL20004-03 CAAAAL20004-03
2-Acetamido-5-aminopyridine 98%
2-Acetamido-5-aminopyridine
Formula: C₇H₉N₃O
Melting Pt: 144…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00160311
CAS Number: 29958-14-3
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR