5-Bromo-2-hydroxy-3-nitropyridine 97%

Danger

Synonyms: 5-Bromo-3-nitro-2-pyridinol

L19850-03 L19850-03 L19850-06 L19850-06
AAAL19850-03EA 28.86 USD
AAAL19850-03 CAAAAL19850-06 AAAL19850-06 CAAAAL19850-03
5-Bromo-2-hydroxy-3-nitropyridine 97%
5-Bromo-2-hydroxy-3-nitropyridine
Formula: C₅H₃BrN₂O₃
Melting Pt: 245…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00023473
CAS Number: 15862-34-7
EINECS: 239-989-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR