5-Chloro-2-hydroxy-3-methylbenzaldehyde 97%

Warning

L19793-06 L19793-03 L19793-06
AAAL19793-06EA 325.48 USD
AAAL19793-06 CAAAAL19793-03 CAAAAL19793-06
5-Chloro-2-hydroxy-3-methylbenzaldehyde 97%
5-Chloro-2-hydroxy-3-methylbenzaldehyde
Formula: C₈H₇ClO₂
MW: 170.60 g/mol
Melting Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00275687
CAS Number: 23602-63-3
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR