6-Chloropicolinic acid 98%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 6-Chloropyridine-2-carboxylic acid2-Chloropyridine-6-carboxylic acid

L19448-06 L19448-03 L19448-06 L19448-MD L19448-03
AAAL19448-03EA 46.1 USD
AAAL19448-03 CAAAAL19448-03 AAAL19448-06 CAAAAL19448-MD CAAAAL19448-06
6-Chloropicolinic acid 98%
6-Chloropicolinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Melting Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00155390
CAS Number: 4684-94-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR