1-Boc-4-hydroxypiperidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-BOC-4-HydroxypiperidineN-Boc-4-piperidinoltert-butyl 4-hydroxypiperidine-1-carboxylatetert-Butyl 4-hydroxypiperidine-1-carboxylate

L19275-06 L19275-03 L19275-06 L19275-03
AAAL19275-03EA 24.16 USD
AAAL19275-03 CAAAAL19275-03 CAAAAL19275-06 AAAL19275-06
1-Boc-4-hydroxypiperidine 98%
1-Boc-4-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD01075174
CAS Number: 109384-19-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR