3-Benzyloxyphenylacetonitrile 97%

Warning

Synonyms: 2-(3-(Benzyloxy)phenyl)acetonitrile

L19241-04 L19241-ME L19241-ME L19241-04
AAAL19241-04EA 137.64 USD
AAAL19241-04 CAAAAL19241-ME CAAAAL19241-04 AAAL19241-ME
3-Benzyloxyphenylacetonitrile 97%
3-Benzyloxyphenylacetonitrile
Formula: C₁₅H₁₃NO
MW: 223.28 g/mol
Boiling Pt: 188…190 °C/3 mm
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00016396
CAS Number: 20967-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR