2,5-Dichloro-3-thenoic acid 98+%

Warning

Synonyms: 2,5-Dichlorothiophene-3-carboxylic acid

L19101-ME L19101-04 L19101-04 L19101-ME
AAAL19101-04EA 146.14 USD
CAAAAL19101-04 AAAL19101-04 CAAAAL19101-ME AAAL19101-ME
2,5-Dichloro-3-thenoic acid 98+%
2,5-Dichloro-3-thenoic acid
Formula: C₅H₂Cl₂O₂S
MW: 197.04 g/mol
Melting Pt: 147…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00082908
CAS Number: 36157-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR