Triclosan 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloro-2-(2, 4-dichlorophenoxy) phenol5-Chloro-2-(2,4-dichlorophenoxy)phenol

L18655-22 L18655-36 L18655-09 L18655-22 L18655-09 L18655-36
AAAL18655-09EA 53.05 USD
AAAL18655-09 CAAAAL18655-09 CAAAAL18655-22 CAAAAL18655-36 AAAL18655-22 AAAL18655-36
Triclosan 99%
Triclosan
Formula: C₁₂H₇Cl₃O₂
MW: 289.54 g/mol
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00800992
CAS Number: 3380-34-5
EINECS: 222-182-2
UN: 3077
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR